Learn More
n-Hexane, 99%, Thermo Scientific Chemicals
£17.70 - £85.40
Chemical Identifiers
CAS | 110-54-3 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392357
|
Thermo Scientific Alfa Aesar
L09938.AD |
50 mL |
£17.70
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302367
|
Thermo Scientific Alfa Aesar
L09938.AK |
250 mL |
£29.20
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312367
|
Thermo Scientific Alfa Aesar
L09938.AU |
1000 mL |
£85.40
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
n-Hexane is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. Other uses are as fuels and fuel additives, intermediates and laboratory chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Hexane is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. Other uses are as fuels and fuel additives, intermediates and laboratory chemicals.
Solubility
Miscible with alcohol, chloroform, acetone and ether.
Notes
Incompatible with dinitrogen tetraoxide. Store in a cool and dry environment.
Chemical Identifiers
110-54-3 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD02179311 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CHEBI:29021 | |
CCCCCC |
Specifications
-95°C | |
0.659 | |
−23°C (−9°F) | |
1.375 | |
MFCD02179311 | |
1730733 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18 | |
n-Hexane |
110-54-3 | |
69°C | |
Weak, Characteristic | |
C6H14 | |
UN1208 | |
14,4694 | |
Miscible with alcohol,chloroform,acetone and ether. | |
CCCCCC | |
86.18 | |
CHEBI:29021 | |
99% |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H335-H336-H361f-H372
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
missing translation for 'einecsNumber' : 203-777-6
missing translation for 'rtecsNumber' : MN9275000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only