missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexane-d14, for NMR, +99.5% atom D, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
Specifications
Specifications
Physical Form | Liquid |
Color | Colorless |
Density | 0.9600g/mL |
Deuterium (D) | 99.50% min. |
Packaging | Glass Bottle |
Quantity | 1 g |
Linear Formula | D3C(CD2)4CD3 |
Formula Weight | 100.28 |
Grade | NMR |
Chemical Name or Material | n-Hexane-d14, 99.5+ atom % D |
Safety and Handling
Product Identifier
- n-Hexane-d14
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction