missing translation for 'onlineSavingsMsg'
Learn More

n-Hexatriacontane, 97+%, Thermo Scientific Chemicals

Product Code. 11354167
Click to view available options
Quantity:
10 g
50 g

n-Hexatriacontane is used as a paraffin wax and in candles. It is used as a key starting material for the preparation of lower alkanes by cracking. It is also used for example in road surfacing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-06-8
Molecular Formula C36H74
Molecular Weight (g/mol) 506.99
MDL Number MFCD00009413
InChI Key YDLYQMBWCWFRAI-UHFFFAOYSA-N
Synonym n-hexatriacontane, hexatriacontane, ch3-ch2 34-ch3, hexatriacontane, analytical standard, hexatriacontane, n, acmc-1b3b7, n-hexatriacontane 10g
PubChem CID 12412
ChEBI CHEBI:72688
IUPAC Name hexatriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 76.2°C
Color White
Density 0.779 g/mL
Boiling Point 497°C
Refractive Index 1.4573
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 506.95
Percent Purity ≥97%
Physical Form Wax
Chemical Name or Material n-Hexatriacontane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'