missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexyllithium, 2.5M (33 wt.%) solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 32.8, 66.8 |
| CAS Max % | 33.2, 67.2 |
| Color | Orange to Yellow |
| Density | 0.6900g/mL |
| Flash Point | −26°C |
| Packaging | AcroSeal™ Glass bottle |
| Linear Formula | LiCH2CH2CH2CH2CH2CH3 |
| Quantity | 100 mL |
| Specific Gravity | 0.69 |
| Solubility Information | Solubility in water: vigorous reaction. |
| Show More |
Safety and Handling
Product Identifier
- n-Hexyllithium, 33 wt.% solution in n-hexane
- Danger
- Flammable liquid Category 2
- Substance which\, in contact with water\, emits flammable gases Category 1
- Pyrophoric liquid Category 1
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H225-Highly flammable liquid and vapour.
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H304-May be fatal if swallowed and enters airways.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P231+P232-Handle under inert gas. Protect from moisture.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrocarbons, C6, n-alkanes, isoalkanes,cyclics, 5-50% n-hexane, n-Hexyllithium
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction