Learn More
N-Hydroxy-2,2-dimethylpropanimidamide, 95%, Thermo Scientific Chemicals
£67.40 - £202.00
Chemical Identifiers
CAS | 42956-75-2 |
---|---|
Molecular Formula | C5H12N2O |
Molecular Weight (g/mol) | 116.16 |
InChI Key | CVOGFMYWFRFWEQ-UHFFFAOYSA-N |
Synonym | n-hydroxypivalimidamide, 1z-n'-hydroxy-2,2-dimethylpropanimidamide, n-hydroxy-2,2-dimethylpropanimidamide, n-hydroxypivalamidine, z-n'-hydroxy-2,2-dimethylpropanimidamide, hydroxydimethylpropanimidamide, z-n'-hydroxypivalimidamide, z-n'-hydroxy-2,2-dimethylpropimidamide |
PubChem CID | 9582836 |
IUPAC Name | N'-hydroxy-2,2-dimethylpropanimidamide |
SMILES | CC(C)(C)C(=NO)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11935671
|
Thermo Scientific Acros
439660010 |
1 g | Glass bottle |
£67.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11925671
|
Thermo Scientific Acros
439660050 |
5 g | Glass bottle |
£202.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
42956-75-2 | |
116.16 | |
n-hydroxypivalimidamide, 1z-n'-hydroxy-2,2-dimethylpropanimidamide, n-hydroxy-2,2-dimethylpropanimidamide, n-hydroxypivalamidine, z-n'-hydroxy-2,2-dimethylpropanimidamide, hydroxydimethylpropanimidamide, z-n'-hydroxypivalimidamide, z-n'-hydroxy-2,2-dimethylpropimidamide | |
N'-hydroxy-2,2-dimethylpropanimidamide |
C5H12N2O | |
CVOGFMYWFRFWEQ-UHFFFAOYSA-N | |
9582836 | |
CC(C)(C)C(=NO)N |
Specifications
42956-75-2 | |
100.0 | |
White to Brown | |
94% min. (HPLC) | |
n-hydroxypivalimidamide, 1z-n'-hydroxy-2,2-dimethylpropanimidamide, n-hydroxy-2,2-dimethylpropanimidamide, n-hydroxypivalamidine, z-n'-hydroxy-2,2-dimethylpropanimidamide, hydroxydimethylpropanimidamide, z-n'-hydroxypivalimidamide, z-n'-hydroxy-2,2-dimethylpropimidamide | |
CC(C)(C)C(=NO)N | |
116.16 | |
116.16 | |
Powder |
94.0 | |
108.6°C to 113.7°C | |
Authentic | |
C5H12N2O | |
CVOGFMYWFRFWEQ-UHFFFAOYSA-N | |
N'-hydroxy-2,2-dimethylpropanimidamide | |
9582836 | |
95% | |
N-Hydroxy-2, 2-dimethylpropanimidamide |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only