missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N'-hydroxy-4-(trifluoromethyl)benzenecarboximidamide, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Spezifikation
Spezifikation
Assay Percent Range | 90% |
Packaging | Amber glass bottle |
Quantity | 5 g |
Formula Weight | 204.15 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | N'-Hydroxy-4-(trifluoromethyl)benzenecarboximidamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'