missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N'-Hydroxypyridine-4-carboximidamide, 97%, Thermo Scientific™
£50.10 - £195.00
Identificadores de productos químicos
CAS | 1594-57-6 |
---|---|
Molecular Formula | C6H7N3O |
Molecular Weight (g/mol) | 137.14 |
MDL Number | MFCD00125873 |
InChI Key | ZUUATXHRJFHSGO-UHFFFAOYSA-N |
Synonym | n'-hydroxypyridine-4-carboximidamide, pyridine-4-amidoxime, 4-pyridylamidoxime, n-hydroxyisonicotinamidine, n-hydroxy-isonicotinamidine, isonicotinamidoxime, isonicotinohydroxamamide, 4-pyridinecarboxamidoxime, n-hydroxyisonicotinimidamide, z-n'-hydroxyisonicotinimidamide |
PubChem CID | 5355713 |
IUPAC Name | nitroso(1H-pyridin-4-ylidene)methanamine |
SMILES | N\C(=N/O)C1=CC=NC=C1 |
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
10566820
|
Thermo Scientific Acros
432480010 |
1 g | Glass bottle | ||||||
10638550
|
Thermo Scientific Acros
432480050 |
5 g | Glass bottle |
£195.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
Identificadores de productos químicos
1594-57-6 | |
137.14 | |
ZUUATXHRJFHSGO-UHFFFAOYSA-N | |
5355713 | |
N\C(=N/O)C1=CC=NC=C1 |
C6H7N3O | |
MFCD00125873 | |
n'-hydroxypyridine-4-carboximidamide, pyridine-4-amidoxime, 4-pyridylamidoxime, n-hydroxyisonicotinamidine, n-hydroxy-isonicotinamidine, isonicotinamidoxime, isonicotinohydroxamamide, 4-pyridinecarboxamidoxime, n-hydroxyisonicotinimidamide, z-n'-hydroxyisonicotinimidamide | |
nitroso(1H-pyridin-4-ylidene)methanamine |
Especificaciones
1594-57-6 | |
100.0 | |
White | |
96% min. (HPLC) | |
MFCD00125873 | |
ZUUATXHRJFHSGO-UHFFFAOYSA-N | |
nitroso(1H-pyridin-4-ylidene)methanamine | |
5355713 | |
97% | |
N'-Hydroxypyridine-4-carboximidamide |
96.0 | |
175.0°C to 180.0°C | |
Authentic | |
C6H7N3O | |
n'-hydroxypyridine-4-carboximidamide, pyridine-4-amidoxime, 4-pyridylamidoxime, n-hydroxyisonicotinamidine, n-hydroxy-isonicotinamidine, isonicotinamidoxime, isonicotinohydroxamamide, 4-pyridinecarboxamidoxime, n-hydroxyisonicotinimidamide, z-n'-hydroxyisonicotinimidamide | |
N\C(=N/O)C1=CC=NC=C1 | |
137.14 | |
137.14 | |
Crystalline Solid |
Seguridad y manipulación
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
Nombre del producto
Palabra de advertencia
Clasificación
Indicaciones de peligro
Consejos de prudencia
- 4-Pyridylamidoxime
Palabra de advertencia
- Warning
Clasificación
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Indicaciones de peligro
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Consejos de prudencia
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only