missing translation for 'onlineSavingsMsg'
Learn More

N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%, Thermo Scientific Chemicals

Product Code. 15400076
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isobutyryl-5′-O-tert-butyldimethylsilyl-2′-deoxyguanosine is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85326-10-9
Molecular Formula C20H33N5O5Si
Molecular Weight (g/mol) 451.599
MDL Number MFCD04972287
InChI Key WZQWRHWLQPEBFF-BFHYXJOUSA-N
Synonym n2-isobutyryl-5'-o-tert-butyldimethylsilyl-2'-deoxyguanosine, n-9-2r,4s,5r-5-tert-butyldimethylsilyl oxy methyl-4-hydroxyoxolan-2-yl-6-oxo-1h-purin-2-yl-2-methylpropanamide
PubChem CID 12016034
IUPAC Name N-[9-[(2R,4S,5R)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide
SMILES CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C3CC(C(O3)CO[Si](C)(C)C(C)(C)C)O

Specifications

Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 451.6
Percent Purity 97%
Chemical Name or Material N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'