missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%, Thermo Scientific Chemicals
£29.70 - £493.00
Chemical Identifiers
CAS | 85326-10-9 |
---|---|
Molecular Formula | C20H33N5O5Si |
Molecular Weight (g/mol) | 451.599 |
MDL Number | MFCD04972287 |
InChI Key | WZQWRHWLQPEBFF-BFHYXJOUSA-N |
Synonym | n2-isobutyryl-5'-o-tert-butyldimethylsilyl-2'-deoxyguanosine, n-9-2r,4s,5r-5-tert-butyldimethylsilyl oxy methyl-4-hydroxyoxolan-2-yl-6-oxo-1h-purin-2-yl-2-methylpropanamide |
PubChem CID | 12016034 |
IUPAC Name | N-[9-[(2R,4S,5R)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide |
SMILES | CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C3CC(C(O3)CO[Si](C)(C)C(C)(C)C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15410076
|
Thermo Scientific Alfa Aesar
H52726.MD |
250 mg | |||||||
15490066
|
Thermo Scientific Alfa Aesar
H52726.03 |
1 g |
£114.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15400076
|
Thermo Scientific Alfa Aesar
H52726.06 |
5 g |
£493.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Isobutyryl-5′-O-tert-butyldimethylsilyl-2′-deoxyguanosine is used as pharmaceutical intermediate.
Notes
Moisture sensitive. Store away from oxidizing agents.
Chemical Identifiers
85326-10-9 | |
451.599 | |
WZQWRHWLQPEBFF-BFHYXJOUSA-N | |
12016034 | |
CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C3CC(C(O3)CO[Si](C)(C)C(C)(C)C)O |
C20H33N5O5Si | |
MFCD04972287 | |
n2-isobutyryl-5'-o-tert-butyldimethylsilyl-2'-deoxyguanosine, n-9-2r,4s,5r-5-tert-butyldimethylsilyl oxy methyl-4-hydroxyoxolan-2-yl-6-oxo-1h-purin-2-yl-2-methylpropanamide | |
N-[9-[(2R,4S,5R)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide |
Specifications
85326-10-9 | |
MFCD04972287 | |
n2-isobutyryl-5'-o-tert-butyldimethylsilyl-2'-deoxyguanosine, n-9-2r,4s,5r-5-tert-butyldimethylsilyl oxy methyl-4-hydroxyoxolan-2-yl-6-oxo-1h-purin-2-yl-2-methylpropanamide | |
CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C3CC(C(O3)CO[Si](C)(C)C(C)(C)C)O | |
451.599 | |
451.6 | |
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine |
C20H33N5O5Si | |
Moisture sensitive | |
WZQWRHWLQPEBFF-BFHYXJOUSA-N | |
N-[9-[(2R,4S,5R)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide | |
12016034 | |
97% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only