Learn More
N-Isopropyl-4-nitroaniline, 97+%, Thermo Scientific™
£24.50 - £270.00
Chemical Identifiers
CAS | 25186-43-0 |
---|---|
Molecular Formula | C9H12N2O2 |
Molecular Weight (g/mol) | 180.207 |
MDL Number | MFCD00128997 |
InChI Key | VTSUWHFLMJLYKN-UHFFFAOYSA-N |
Synonym | n-isopropyl-4-nitroaniline, benzenamine, n-1-methylethyl-4-nitro, 4-isopropylamino-nitrobenzene, p-nitrophenyl-isopropylamine, 4-nitro-n-propan-2-yl aniline, n-1-methylethyl-4-nitroaniline, benzenamine,n-1-methylethyl-4-nitro |
PubChem CID | 32833 |
IUPAC Name | 4-nitro-N-propan-2-ylaniline |
SMILES | CC(C)NC1=CC=C(C=C1)[N+](=O)[O-] |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
15499067
|
Thermo Scientific Alfa Aesar
H52375.MD |
250 mg | |||||||
15479067
|
Thermo Scientific Alfa Aesar
H52375.03 |
1 g | |||||||
15489067
|
Thermo Scientific Alfa Aesar
H52375.06 |
5 g |
£270.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
25186-43-0 | |
180.207 | |
VTSUWHFLMJLYKN-UHFFFAOYSA-N | |
32833 | |
CC(C)NC1=CC=C(C=C1)[N+](=O)[O-] |
C9H12N2O2 | |
MFCD00128997 | |
n-isopropyl-4-nitroaniline, benzenamine, n-1-methylethyl-4-nitro, 4-isopropylamino-nitrobenzene, p-nitrophenyl-isopropylamine, 4-nitro-n-propan-2-yl aniline, n-1-methylethyl-4-nitroaniline, benzenamine,n-1-methylethyl-4-nitro | |
4-nitro-N-propan-2-ylaniline |
Spécification
25186-43-0 | |
MFCD00128997 | |
n-isopropyl-4-nitroaniline, benzenamine, n-1-methylethyl-4-nitro, 4-isopropylamino-nitrobenzene, p-nitrophenyl-isopropylamine, 4-nitro-n-propan-2-yl aniline, n-1-methylethyl-4-nitroaniline, benzenamine,n-1-methylethyl-4-nitro | |
CC(C)NC1=CC=C(C=C1)[N+](=O)[O-] | |
180.207 | |
180.2 | |
N-Isopropyl-4-nitroaniline |
C9H12N2O2 | |
UN2811 | |
VTSUWHFLMJLYKN-UHFFFAOYSA-N | |
4-nitro-N-propan-2-ylaniline | |
32833 | |
≥97% |
Sécurité et traitement
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 246-721-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- N-Isopropyl-4-nitroaniline
Mention d'avertissement
- Warning
Catégorie de danger
- Acute toxicity Category 4
Mention de danger
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
Conseils de prudence
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Informations supplémentaires
- MIXTURE LIST-Contains : N-(1-methylethyl)-4-nitrobenzenamine