missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropyl-N'-phenyl-p-phenylenediamine, 95%, Thermo Scientific Chemicals

£25.40 - £79.10

Chemical Identifiers

CAS 101-72-4
Molecular Formula C15H18N2
Molecular Weight (g/mol) 226.323
MDL Number MFCD00026348
InChI Key OUBMGJOQLXMSNT-UHFFFAOYSA-N
Synonym ippd, n-isopropyl-n'-phenyl-p-phenylenediamine, n-isopropyl-n'-phenyl-1,4-phenylenediamine, cyzone, 4-anilino-n-isopropylaniline, antigen 3c, antioxidant ip, santoflex ip, diaphen fp, cyzone ip
PubChem CID 7573
ChEBI CHEBI:63569
IUPAC Name 1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine
SMILES CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11477056
View Documents
Thermo Scientific Alfa Aesar
B25378.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11467056
View Documents
Thermo Scientific Alfa Aesar
B25378.A1
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

101-72-4
226.323
OUBMGJOQLXMSNT-UHFFFAOYSA-N
7573
1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine
C15H18N2
MFCD00026348
ippd, n-isopropyl-n'-phenyl-p-phenylenediamine, n-isopropyl-n'-phenyl-1,4-phenylenediamine, cyzone, 4-anilino-n-isopropylaniline, antigen 3c, antioxidant ip, santoflex ip, diaphen fp, cyzone ip
CHEBI:63569
CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

101-72-4
1.1
172°C (342°F)
MFCD00026348
Air Sensitive
OUBMGJOQLXMSNT-UHFFFAOYSA-N
1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine
7573
226.32
N-Isopropyl-N'-phenyl-p-phenylenediamine
74°C to 80°C
161°C (1 mmHg)
C15H18N2
UN3077
ippd, n-isopropyl-n'-phenyl-p-phenylenediamine, n-isopropyl-n'-phenyl-1,4-phenylenediamine, cyzone, 4-anilino-n-isopropylaniline, antigen 3c, antioxidant ip, santoflex ip, diaphen fp, cyzone ip
CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2
226.323
CHEBI:63569
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 202-969-7

missing translation for 'rtecsNumber' : ST2650000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only