missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylmethylamine, 98%, Thermo Scientific Chemicals

Product Code. 11985181
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4747-21-1
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
InChI Key XHFGWHUWQXTGAT-UHFFFAOYSA-N
Synonym n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine
PubChem CID 78485
IUPAC Name N-methylpropan-2-amine
SMILES CC(C)NC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.7020g/mL
Boiling Point 50°C to 53°C
Flash Point -32°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3NHCH(CH3)2
Refractive Index 1.3830 to 1.3850 (20°C, 589nm)
Quantity 50 mL
Specific Gravity 0.702
Formula Weight 73.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Isopropylmethylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Isopropylmethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'