missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1,2-phenylenediamine, 97%, Thermo Scientific Chemicals

£81.90 - £270.00

Chemical Identifiers

CAS 4760-34-3
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00192314
InChI Key RPKCLSMBVQLWIN-UHFFFAOYSA-N
Synonym n1-methylbenzene-1,2-diamine, n-methyl-o-phenylenediamine, n-methylbenzene-1,2-diamine, 1,2-benzenediamine, n-methyl, n-methyl-1,2-phenylenediamine, n-methyl-1,2-benzenediamine, 2-amino-n-methylaniline, n-methyl-2-phenylenediamine, 2-methylamino aniline, unii-roi30fpr50
PubChem CID 78498
IUPAC Name 2-N-methylbenzene-1,2-diamine
SMILES CNC1=CC=CC=C1N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10618360
View Documents Promotion Details
Thermo Scientific Acros
434230050
5 mL Glass bottle
£81.90
5mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10311670
View Documents Promotion Details
Thermo Scientific Acros
434230250
25 mL Glass bottle
£270.00
25mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4760-34-3
122.17
RPKCLSMBVQLWIN-UHFFFAOYSA-N
78498
CNC1=CC=CC=C1N
C7H10N2
MFCD00192314
n1-methylbenzene-1,2-diamine, n-methyl-o-phenylenediamine, n-methylbenzene-1,2-diamine, 1,2-benzenediamine, n-methyl, n-methyl-1,2-phenylenediamine, n-methyl-1,2-benzenediamine, 2-amino-n-methylaniline, n-methyl-2-phenylenediamine, 2-methylamino aniline, unii-roi30fpr50
2-N-methylbenzene-1,2-diamine

Specifications

4760-34-3
100.0
Brown to Yellow
123°C to 124°C (10.0 mmHg)
Authentic
C7H10N2
1.075
RPKCLSMBVQLWIN-UHFFFAOYSA-N
2-N-methylbenzene-1,2-diamine
78498
97%
N-Methyl-1, 2-phenylenediamine
96.0
22°C
1.0750g/mL
113°C
96% min. (GC)
MFCD00192314
n1-methylbenzene-1,2-diamine, n-methyl-o-phenylenediamine, n-methylbenzene-1,2-diamine, 1,2-benzenediamine, n-methyl, n-methyl-1,2-phenylenediamine, n-methyl-1,2-benzenediamine, 2-amino-n-methylaniline, n-methyl-2-phenylenediamine, 2-methylamino aniline, unii-roi30fpr50
CNC1=CC=CC=C1N
122.17
122.17
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 225-297-6

Product Identifier
  • N-Methyl-1,2-phenylenediamine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only