Learn More
N-Methyl-(1-methylindolin-5-yl)methylamine, 90%, Thermo Scientific™
£80.55 - £219.00
Chemical Identifiers
CAS | 906352-81-6 |
---|---|
Molecular Formula | C11H16N2 |
Molecular Weight (g/mol) | 176.26 |
MDL Number | MFCD09817499 |
InChI Key | QPUPWFNEDMKWGL-UHFFFAOYSA-N |
Synonym | 1-methyl-5-methylamino methyl indoline, 2,3-dihydro-1-methyl-5-methylamino methyl-1h-indole, methyl 1-methyl-2,3-dihydroindol-5-yl methyl amine, methyl 1-methyl-2,3-dihydro-1h-indol-5-yl methyl amine, methyl 1-methylindolin-5-yl methyl amine, n-methyl-1-1-methylindolin-5-yl methanamine, 1-2,3-dihydro-1-methyl-1h-indol-5-yl-n-methylmethylamine, n-methyl-1-1-methyl-2,3-dihydro-1h-indol-5-yl methanamine |
PubChem CID | 24229592 |
IUPAC Name | N-methyl-1-(1-methyl-2,3-dihydroindol-5-yl)methanamine |
SMILES | CNCC1=CC2=C(C=C1)N(C)CC2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10376664
|
Thermo Scientific Maybridge
CC58746CB |
250 mg | Amber Glass Bottle |
£80.55
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10692665
|
Thermo Scientific Maybridge
CC58746DA |
1 g | Amber Glass Bottle |
£219.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
906352-81-6 | |
176.26 | |
QPUPWFNEDMKWGL-UHFFFAOYSA-N | |
24229592 | |
CNCC1=CC2=C(C=C1)N(C)CC2 |
C11H16N2 | |
MFCD09817499 | |
1-methyl-5-methylamino methyl indoline, 2,3-dihydro-1-methyl-5-methylamino methyl-1h-indole, methyl 1-methyl-2,3-dihydroindol-5-yl methyl amine, methyl 1-methyl-2,3-dihydro-1h-indol-5-yl methyl amine, methyl 1-methylindolin-5-yl methyl amine, n-methyl-1-1-methylindolin-5-yl methanamine, 1-2,3-dihydro-1-methyl-1h-indol-5-yl-n-methylmethylamine, n-methyl-1-1-methyl-2,3-dihydro-1h-indol-5-yl methanamine | |
N-methyl-1-(1-methyl-2,3-dihydroindol-5-yl)methanamine |
Specifications
906352-81-6 | |
100.0 | |
C11H16N2 | |
1-methyl-5-methylamino methyl indoline, 2,3-dihydro-1-methyl-5-methylamino methyl-1h-indole, methyl 1-methyl-2,3-dihydroindol-5-yl methyl amine, methyl 1-methyl-2,3-dihydro-1h-indol-5-yl methyl amine, methyl 1-methylindolin-5-yl methyl amine, n-methyl-1-1-methylindolin-5-yl methanamine, 1-2,3-dihydro-1-methyl-1h-indol-5-yl-n-methylmethylamine, n-methyl-1-1-methyl-2,3-dihydro-1h-indol-5-yl methanamine | |
CNCC1=CC2=C(C=C1)N(C)CC2 | |
176.26 | |
176.26 | |
N-methyl-(1-methylindolin-5-yl)methylamine |
89.5 | |
90% | |
MFCD09817499 | |
QPUPWFNEDMKWGL-UHFFFAOYSA-N | |
N-methyl-1-(1-methyl-2,3-dihydroindol-5-yl)methanamine | |
24229592 | |
90% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger