missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[(2,3-dihydrobenzo[b]furan-7-yl)methyl]amine, 90%, Thermo Scientific™

£47.20 - £128.00

Chemical Identifiers

CAS 389845-43-6
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD10700046
InChI Key WVSLYVGYBIMHJY-UHFFFAOYSA-N
Synonym n-methyl-2,3-dihydrobenzo b furan-7-yl methyl amine, 2,3-dihydro-1-benzofuran-7-ylmethyl methyl amine, 1-2,3-dihydrobenzofuran-7-yl-n-methylmethanamine, 2,3-dihydro-benzofuran-7-ylmethyl methylamine, 7-benzofuranmethanamine,2,3-dihydro-n-methyl, 2,3-dihydrobenzo 2,3-b furan-7-ylmethyl methylamine, 1-2,3-dihydro-1-benzofuran-7-yl-n-methylmethanamine
PubChem CID 26343571
IUPAC Name 1-(2,3-dihydro-1-benzofuran-7-yl)-N-methylmethanamine
SMILES CNCC1=CC=CC2=C1OCC2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10142914
View Documents Promotion Details
Thermo Scientific Maybridge
CC00946CB
250 mg Amber Glass Bottle
£47.20
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10633985
View Documents Promotion Details
Thermo Scientific Maybridge
CC00946DA
1 g Amber Glass Bottle
£128.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

389845-43-6
163.22
WVSLYVGYBIMHJY-UHFFFAOYSA-N
26343571
CNCC1=CC=CC2=C1OCC2
C10H13NO
MFCD10700046
n-methyl-2,3-dihydrobenzo b furan-7-yl methyl amine, 2,3-dihydro-1-benzofuran-7-ylmethyl methyl amine, 1-2,3-dihydrobenzofuran-7-yl-n-methylmethanamine, 2,3-dihydro-benzofuran-7-ylmethyl methylamine, 7-benzofuranmethanamine,2,3-dihydro-n-methyl, 2,3-dihydrobenzo 2,3-b furan-7-ylmethyl methylamine, 1-2,3-dihydro-1-benzofuran-7-yl-n-methylmethanamine
1-(2,3-dihydro-1-benzofuran-7-yl)-N-methylmethanamine

Specifications

389845-43-6
100.0
C10H13NO
n-methyl-2,3-dihydrobenzo b furan-7-yl methyl amine, 2,3-dihydro-1-benzofuran-7-ylmethyl methyl amine, 1-2,3-dihydrobenzofuran-7-yl-n-methylmethanamine, 2,3-dihydro-benzofuran-7-ylmethyl methylamine, 7-benzofuranmethanamine,2,3-dihydro-n-methyl, 2,3-dihydrobenzo 2,3-b furan-7-ylmethyl methylamine, 1-2,3-dihydro-1-benzofuran-7-yl-n-methylmethanamine
CNCC1=CC=CC2=C1OCC2
163.22
163.22
N-methyl-[(2, 3-dihydrobenzo[b]furan-7-yl)methyl]amine
89.5
90%
MFCD10700046
WVSLYVGYBIMHJY-UHFFFAOYSA-N
1-(2,3-dihydro-1-benzofuran-7-yl)-N-methylmethanamine
26343571
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications