Learn More
N-Methyl-2-nitroaniline, 98%, Thermo Scientific Chemicals
£39.00 - £501.00
Chemical Identifiers
CAS | 612-28-2 |
---|---|
Molecular Formula | C7H8N2O2 |
Molecular Weight (g/mol) | 152.15 |
MDL Number | MFCD00007090 |
InChI Key | KFBOUJZFFJDYTA-UHFFFAOYSA-N |
Synonym | n-methyl-o-nitroaniline, benzenamine, n-methyl-2-nitro, n-methyl 2-nitroaniline, o-methylamino nitrobenzene, aniline, n-methyl-o-nitro, 2-nitro-n-methylaniline, o-nitro-n-methylaniline, n-methyl-2-nitrobenzenamine, o-nitro-n-methyl aniline, methyl 2-nitrophenyl amine |
PubChem CID | 69157 |
IUPAC Name | N-methyl-2-nitroaniline |
SMILES | CNC1=CC=CC=C1[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11464107
|
Thermo Scientific Alfa Aesar
A19538.06 |
5 g |
£39.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474107
|
Thermo Scientific Alfa Aesar
A19538.14 |
25 g |
£148.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484107
|
Thermo Scientific Alfa Aesar
A19538.22 |
100 g |
£501.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
612-28-2 | |
152.15 | |
KFBOUJZFFJDYTA-UHFFFAOYSA-N | |
69157 | |
CNC1=CC=CC=C1[N+]([O-])=O |
C7H8N2O2 | |
MFCD00007090 | |
n-methyl-o-nitroaniline, benzenamine, n-methyl-2-nitro, n-methyl 2-nitroaniline, o-methylamino nitrobenzene, aniline, n-methyl-o-nitro, 2-nitro-n-methylaniline, o-nitro-n-methylaniline, n-methyl-2-nitrobenzenamine, o-nitro-n-methyl aniline, methyl 2-nitrophenyl amine | |
N-methyl-2-nitroaniline |
Specifications
612-28-2 | |
>110°C (230°F) | |
C7H8N2O2 | |
UN2811 | |
KFBOUJZFFJDYTA-UHFFFAOYSA-N | |
N-methyl-2-nitroaniline | |
69157 | |
98% |
33°C to 36°C | |
Characteristic | |
MFCD00007090 | |
n-methyl-o-nitroaniline, benzenamine, n-methyl-2-nitro, n-methyl 2-nitroaniline, o-methylamino nitrobenzene, aniline, n-methyl-o-nitro, 2-nitro-n-methylaniline, o-nitro-n-methylaniline, n-methyl-2-nitrobenzenamine, o-nitro-n-methyl aniline, methyl 2-nitrophenyl amine | |
CNC1=CC=CC=C1[N+]([O-])=O | |
152.15 | |
152.15 | |
N-Methyl-2-nitroaniline |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 210-303-1
missing translation for 'rtecsNumber' : BY5710000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only