missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-nitroaniline, 97%, Thermo Scientific Chemicals

Product Code. 15481358
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-26-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00963641
InChI Key IKSRCCUOUJJGAU-UHFFFAOYSA-N
Synonym 3-nitro-n-methylaniline, n-methyl-m-nitroaniline, benzenamine, n-methyl-3-nitro, n-methyl-3-nitro-aniline, acmc-1b45l, n-methyl-n-3-nitrophenyl amine #
PubChem CID 219622
IUPAC Name N-methyl-3-nitroaniline
SMILES CNC1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 64°C to 68°C
Color White to Brown
Quantity 1 g
Formula Weight 152.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Methyl-3-nitroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'