missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-pyrimidin-2-ylbenzylamine, 97%, Thermo Scientific™

£50.60 - £108.00

Chemical Identifiers

CAS 886851-49-6
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.257
MDL Number MFCD09702386
InChI Key AHOVMAYXXRVWIS-UHFFFAOYSA-N
Synonym n-methyl-3-pyrimidin-2-ylbenzylamine, methyl 3-pyrimidin-2-yl phenyl methyl amine, benzenemethanamine,n-methyl-3-2-pyrimidinyl, n-methyl-1-3-pyrimidin-2-yl phenyl methanamine, n-3-pyrimidin-2-yl benzyl methylamine, 2-3-methylamino methyl phenyl pyrimidine, methyl 3-pyrimidin-2-ylphenyl methyl amine, benzenemethanamine, n-methyl-3-2-pyrimidinyl-9ci
PubChem CID 24229568
IUPAC Name N-methyl-1-(3-pyrimidin-2-ylphenyl)methanamine
SMILES CNCC1=CC=CC(=C1)C2=NC=CC=N2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10357014
View Documents Promotion Details
Thermo Scientific Maybridge
CC56346CB
250 mg Amber Glass Bottle
£50.60
250mg
Please to purchase this item. Need a web account? Register with us today!
10183374
View Documents Promotion Details
Thermo Scientific Maybridge
CC56346DA
1 g Amber Glass Bottle
£108.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

886851-49-6
199.257
AHOVMAYXXRVWIS-UHFFFAOYSA-N
24229568
CNCC1=CC=CC(=C1)C2=NC=CC=N2
C12H13N3
MFCD09702386
n-methyl-3-pyrimidin-2-ylbenzylamine, methyl 3-pyrimidin-2-yl phenyl methyl amine, benzenemethanamine,n-methyl-3-2-pyrimidinyl, n-methyl-1-3-pyrimidin-2-yl phenyl methanamine, n-3-pyrimidin-2-yl benzyl methylamine, 2-3-methylamino methyl phenyl pyrimidine, methyl 3-pyrimidin-2-ylphenyl methyl amine, benzenemethanamine, n-methyl-3-2-pyrimidinyl-9ci
N-methyl-1-(3-pyrimidin-2-ylphenyl)methanamine

Specifications

886851-49-6
100.0
C12H13N3
n-methyl-3-pyrimidin-2-ylbenzylamine, methyl 3-pyrimidin-2-yl phenyl methyl amine, benzenemethanamine,n-methyl-3-2-pyrimidinyl, n-methyl-1-3-pyrimidin-2-yl phenyl methanamine, n-3-pyrimidin-2-yl benzyl methylamine, 2-3-methylamino methyl phenyl pyrimidine, methyl 3-pyrimidin-2-ylphenyl methyl amine, benzenemethanamine, n-methyl-3-2-pyrimidinyl-9ci
CNCC1=CC=CC(=C1)C2=NC=CC=N2
199.257
199.26
N-methyl-3-pyrimidin-2-ylbenzylamine
96.5
97%
MFCD09702386
AHOVMAYXXRVWIS-UHFFFAOYSA-N
N-methyl-1-(3-pyrimidin-2-ylphenyl)methanamine
24229568
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications