missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-{[5-(morpholinomethyl)-2-furyl]methyl}amine, 97%, Thermo Scientific™

Product Code. 10778245
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 893741-66-7
Molecular Formula C11H18N2O2
Molecular Weight (g/mol) 210.28
MDL Number MFCD06803236
InChI Key RVGSMPQAXGUMNM-UHFFFAOYSA-N
Synonym n-methyl-5-morpholinomethyl-2-furyl methyl amine, methyl 5-morpholin-4-ylmethyl furan-2-yl methyl amine, n-methyl-1-5-morpholin-4-ylmethyl furan-2-yl methylamine, 2-furanmethanamine,n-methyl-5-4-morpholinylmethyl, 4-5-methylamino methyl furan-2-ylmethyl morpholine, methyl 5-morpholin-4-ylmethyl 2-furyl methyl amine, n-methyl-1-5-morpholinomethyl furan-2-yl methanamine, n-methyl-1-5-morpholin-4-yl methyl furan-2-yl methanamine
PubChem CID 16495000
IUPAC Name N-methyl-1-[5-(morpholin-4-ylmethyl)furan-2-yl]methanamine
SMILES CNCC1=CC=C(CN2CCOCC2)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 210.28
Percent Purity 97%
Chemical Name or Material N-methyl-{[5-(morpholinomethyl)-2-furyl]methyl}amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'