Learn More
n-methyl-(5-pyrid-3-ylthien-2-yl)methylamine, 97%, Thermo Scientific™
£42.60 - £106.00
Chemical Identifiers
CAS | 837376-49-5 |
---|---|
Molecular Formula | C11H12N2S |
Molecular Weight (g/mol) | 204.291 |
MDL Number | MFCD11109328 |
InChI Key | ZSTXLHXHAPANSL-UHFFFAOYSA-N |
Synonym | methyl 5-pyridin-3-yl thiophen-2-yl methyl amine, n-methyl-5-pyrid-3-ylthien-2-yl methylamine, n-methyl-5-pyridin-3-yl thiophen-2-yl methylamine, 2-methylamino methyl-5-pyridin-3-ylthiophene, n-methyl-1-5-pyridin-3-yl thiophen-2-yl methanamine, nicotine 3-heteroaromatic analogue 1b, methyl 5-3-pyridyl 2-thienyl methyl amine, 2-thiophenemethanamine,n-methyl-5-3-pyridinyl, 2-thiophenemethanamine, n-methyl-5-3-pyridinyl |
PubChem CID | 11321674 |
IUPAC Name | N-methyl-1-(5-pyridin-3-ylthiophen-2-yl)methanamine |
SMILES | CNCC1=CC=C(S1)C2=CN=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10567294
|
Thermo Scientific Maybridge
CC66846CB |
250 mg | Amber Glass Bottle |
£42.60
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10653265
|
Thermo Scientific Maybridge
CC66846DA |
1 g | Amber Glass Bottle |
£106.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
837376-49-5 | |
204.291 | |
ZSTXLHXHAPANSL-UHFFFAOYSA-N | |
11321674 | |
CNCC1=CC=C(S1)C2=CN=CC=C2 |
C11H12N2S | |
MFCD11109328 | |
methyl 5-pyridin-3-yl thiophen-2-yl methyl amine, n-methyl-5-pyrid-3-ylthien-2-yl methylamine, n-methyl-5-pyridin-3-yl thiophen-2-yl methylamine, 2-methylamino methyl-5-pyridin-3-ylthiophene, n-methyl-1-5-pyridin-3-yl thiophen-2-yl methanamine, nicotine 3-heteroaromatic analogue 1b, methyl 5-3-pyridyl 2-thienyl methyl amine, 2-thiophenemethanamine,n-methyl-5-3-pyridinyl, 2-thiophenemethanamine, n-methyl-5-3-pyridinyl | |
N-methyl-1-(5-pyridin-3-ylthiophen-2-yl)methanamine |
Specifications
837376-49-5 | |
100.0 | |
C11H12N2S | |
methyl 5-pyridin-3-yl thiophen-2-yl methyl amine, n-methyl-5-pyrid-3-ylthien-2-yl methylamine, n-methyl-5-pyridin-3-yl thiophen-2-yl methylamine, 2-methylamino methyl-5-pyridin-3-ylthiophene, n-methyl-1-5-pyridin-3-yl thiophen-2-yl methanamine, nicotine 3-heteroaromatic analogue 1b, methyl 5-3-pyridyl 2-thienyl methyl amine, 2-thiophenemethanamine,n-methyl-5-3-pyridinyl, 2-thiophenemethanamine, n-methyl-5-3-pyridinyl | |
CNCC1=CC=C(S1)C2=CN=CC=C2 | |
204.291 | |
204.3 | |
N-methyl-(5-pyrid-3-ylthien-2-yl)methylamine |
96.5 | |
97% | |
MFCD11109328 | |
ZSTXLHXHAPANSL-UHFFFAOYSA-N | |
N-methyl-1-(5-pyridin-3-ylthiophen-2-yl)methanamine | |
11321674 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger