missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Methyl-beta-alaninenitrile, 98%

Product Code. 10513101
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-05-0
Molecular Formula C4H8N2
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001954
InChI Key UNIJBMUBHBAUET-UHFFFAOYSA-N
Synonym 3-methylamino propanenitrile, 3-methylaminopropionitrile, 3-methylamino propionitrile, propanenitrile, 3-methylamino, 2-cyanoethyl methyl amine, propionitrile, 3-methylamino, n-2-cyanoethyl methylamine, 3-n-methylamino propionitrile, 2-cyanoethyl methylamine, n-methyl-beta-alaninenitrile
PubChem CID 69656
IUPAC Name 3-(methylamino)propanenitrile
SMILES CNCCC#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 0.8900g/mL
Boiling Point 182°C to 186°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3NHCH2CH2CN
Refractive Index 1.4300 to 1.432
Quantity 50 g
Specific Gravity 0.89
Formula Weight 84.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Methyl-β-alaninenitrile, 98%
Show More Show Less
compliance-icons
Product Identifier
  • N-Methyl-ß-alaninenitrile
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'