missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-N-(2-phenoxybenzyl)amine, Thermo Scientific™

Product Code. 10596975
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 361394-74-3
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
InChI Key IMIUMEQDSKHKST-UHFFFAOYSA-N
Synonym n-methyl-2-phenoxybenzylamine, n-methyl-1-2-phenoxyphenyl methanamine, n-methyl-n-2-phenoxybenzyl amine, methyl 2-phenoxyphenyl methyl amine, benzenemethanamine, n-methyl-2-phenoxy, n-methyl-2-phenoxybenzenemethanamine, n-methyl-n-2-phenoxy benzylamine, n-methyl-1-2-phenoxyphenyl ;methanamine, n-methyl-1-2-phenoxyphenyl methanamine hydrochloride
PubChem CID 23512750
IUPAC Name N-methyl-1-(2-phenoxyphenyl)methanamine
SMILES CNCC1=CC=CC=C1OC2=CC=CC=C2

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 213.28
Percent Purity 95%
Chemical Name or Material N-Methyl-N-(2-phenoxybenzyl)amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.