missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-methyl-N-(2-phenoxybenzyl)amine, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 95% |
Quantity | 1 g |
Formula Weight | 213.28 |
Percent Purity | 95% |
Chemical Name or Material | N-Methyl-N-(2-phenoxybenzyl)amine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction