missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-N-(2-phenoxybenzyl)amine, Thermo Scientific™

£23.00 - £320.00

Chemical Identifiers

CAS 361394-74-3
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
InChI Key IMIUMEQDSKHKST-UHFFFAOYSA-N
Synonym n-methyl-2-phenoxybenzylamine, n-methyl-1-2-phenoxyphenyl methanamine, n-methyl-n-2-phenoxybenzyl amine, methyl 2-phenoxyphenyl methyl amine, benzenemethanamine, n-methyl-2-phenoxy, n-methyl-2-phenoxybenzenemethanamine, n-methyl-n-2-phenoxy benzylamine, n-methyl-1-2-phenoxyphenyl ;methanamine, n-methyl-1-2-phenoxyphenyl methanamine hydrochloride
PubChem CID 23512750
IUPAC Name N-methyl-1-(2-phenoxyphenyl)methanamine
SMILES CNCC1=CC=CC=C1OC2=CC=CC=C2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10205914
View Documents
Thermo Scientific Maybridge
CC49946CB
250 mg
£23.00
250mg
Please to purchase this item. Need a web account? Register with us today!
10772295
View Documents
Thermo Scientific Maybridge
CC49946DA
1 g
£78.20
1g
Please to purchase this item. Need a web account? Register with us today!
10596975
View Documents
Thermo Scientific Maybridge
CC49946DE
5 g
£320.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

361394-74-3
213.28
n-methyl-2-phenoxybenzylamine, n-methyl-1-2-phenoxyphenyl methanamine, n-methyl-n-2-phenoxybenzyl amine, methyl 2-phenoxyphenyl methyl amine, benzenemethanamine, n-methyl-2-phenoxy, n-methyl-2-phenoxybenzenemethanamine, n-methyl-n-2-phenoxy benzylamine, n-methyl-1-2-phenoxyphenyl ;methanamine, n-methyl-1-2-phenoxyphenyl methanamine hydrochloride
N-methyl-1-(2-phenoxyphenyl)methanamine
C14H15NO
IMIUMEQDSKHKST-UHFFFAOYSA-N
23512750
CNCC1=CC=CC=C1OC2=CC=CC=C2

Specifications

361394-74-3
C14H15NO
IMIUMEQDSKHKST-UHFFFAOYSA-N
N-methyl-1-(2-phenoxyphenyl)methanamine
23512750
95%
95%
n-methyl-2-phenoxybenzylamine, n-methyl-1-2-phenoxyphenyl methanamine, n-methyl-n-2-phenoxybenzyl amine, methyl 2-phenoxyphenyl methyl amine, benzenemethanamine, n-methyl-2-phenoxy, n-methyl-2-phenoxybenzenemethanamine, n-methyl-n-2-phenoxy benzylamine, n-methyl-1-2-phenoxyphenyl ;methanamine, n-methyl-1-2-phenoxyphenyl methanamine hydrochloride
CNCC1=CC=CC=C1OC2=CC=CC=C2
213.28
213.28
N-Methyl-N-(2-phenoxybenzyl)amine
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.