missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(5-methyl-3-phenylisoxazol-4-yl)methyl]amine, 97%, Thermo Scientific™

Product Code. 10361573
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-57-9
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.26
MDL Number MFCD07772789
InChI Key IEMWBRRLWLUKQI-UHFFFAOYSA-N
Synonym n-methyl-n-5-methyl-3-phenylisoxazol-4-yl methyl amine, 4-isoxazolemethanamine,n,5-dimethyl-3-phenyl, methyl 5-methyl-3-phenyl-1,2-oxazol-4-yl methyl amine, n-methyl-1-5-methyl-3-phenyl-1,2-oxazol-4-yl methanamine, methyl 5-methyl-3-phenylisoxazol-4-yl methyl amine
PubChem CID 7162097
IUPAC Name N-methyl-1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)methanamine
SMILES CNCC1=C(C)ON=C1C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material N-methyl-N-[(5-methyl-3-phenylisoxazol-4-yl)methyl]amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'