missing translation for 'onlineSavingsMsg'
Learn More
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%, Thermo Scientific Chemicals Product Code.: 10226850

N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%, Thermo Scientific Chemicals

Product Code. 10226850
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10226850

Brand: Thermo Scientific Acros 221580050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent, Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24589-78-4
Molecular Formula C6H12F3NOSi
Molecular Weight (g/mol) 199.25
MDL Number MFCD00000411
InChI Key MSPCIZMDDUQPGJ-UHFFFAOYSA-N
Synonym mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm
PubChem CID 32510
ChEBI CHEBI:85064
IUPAC Name 2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide
SMILES CN(C(=O)C(F)(F)F)[Si](C)(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 130°C to 132°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CF3CON(CH3)Si(CH3)3
Refractive Index 1.3782 to 1.3802
Quantity 5 g
Specific Gravity 1.07
Solubility Information Solubility in water: reacts
Formula Weight 199.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.