missing translation for 'onlineSavingsMsg'
Learn More

N-Methylaniline, 98%, Thermo Scientific Chemicals

£14.40 - £84.70

Chemical Identifiers

CAS 100-61-8
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00008283
InChI Key AFBPFSWMIHJQDM-UHFFFAOYSA-N
Synonym methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
PubChem CID 7515
ChEBI CHEBI:15733
IUPAC Name N-methylaniline
SMILES CNC1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11313297
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L13873.22
100 g
£14.40
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11323297
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L13873.36
500 g
£29.10
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11333297
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L13873.0E
2500 g
£84.70
2.50kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N-Methylaniline is used in the preparation of N,N'-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylaniline is used in the preparation of N,N′-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides, acid chlorides and carbon dioxide.
Specifications

Chemical Identifiers

100-61-8
107.16
AFBPFSWMIHJQDM-UHFFFAOYSA-N
7515
N-methylaniline
C7H9N
MFCD00008283
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
CHEBI:15733
CNC1=CC=CC=C1

Specifications

100-61-8
0.989
86°C (186°F)
1.571
UN2294
Air Sensitive
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
AFBPFSWMIHJQDM-UHFFFAOYSA-N
N-methylaniline
7515
107.16
N-Methylaniline
-57°C
194°C to 197°C
C7H9N
MFCD00008283
741982
14,6019
Immiscible with water.
CNC1=CC=CC=C1
107.16
CHEBI:15733
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c

H227-H301+H311+H331-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N-METHYLANILINE

missing translation for 'einecsNumber' : 202-870-9

missing translation for 'rtecsNumber' : BY4550000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only