Learn More
N-Methylethylenediamine, 95%, Thermo Scientific Chemicals
£39.80 - £356.78
Chemical Identifiers
CAS | 109-81-9 |
---|---|
Molecular Formula | C3H12N2 |
Molecular Weight (g/mol) | 76.14 |
MDL Number | MFCD00008165 |
InChI Key | KFIGICHILYTCJF-UHFFFAOYSA-P |
Synonym | n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine |
PubChem CID | 8014 |
IUPAC Name | N'-methylethane-1,2-diamine |
SMILES | C[NH2+]CC[NH3+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474793
|
Thermo Scientific Alfa Aesar
A12165.06 |
5 g |
£39.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484793
|
Thermo Scientific Alfa Aesar
A12165.14 |
25 g |
£120.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494793
|
Thermo Scientific Alfa Aesar
A12165.22 |
100 g |
£356.78
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Methylethylenediamine undergoes condensation with pyridine-2-carboxaldehyde to form an imine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Methylethylenediamine undergoes condensation with pyridine-2-carboxaldehyde to form an imine.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Chemical Identifiers
109-81-9 | |
76.14 | |
KFIGICHILYTCJF-UHFFFAOYSA-P | |
8014 | |
C[NH2+]CC[NH3+] |
C3H12N2 | |
MFCD00008165 | |
n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine | |
N'-methylethane-1,2-diamine |
Specifications
109-81-9 | |
114°C to 116°C | |
Amine-like | |
H2NCH2CH2NHCH3 | |
MFCD00008165 | |
878141 | |
n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine | |
C[NH2+]CC[NH3+] | |
76.14 | |
74.13 | |
N-Methylethylenediamine |
0.851 | |
41°C (105°F) | |
C3H12N2 | |
1.4395 | |
UN2734 | |
Air sensitive; Hygroscopic | |
KFIGICHILYTCJF-UHFFFAOYSA-P | |
N'-methylethane-1,2-diamine | |
8014 | |
95% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 203-707-4
missing translation for 'rtecsNumber' : KV5250500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only