missing translation for 'onlineSavingsMsg'
Learn More
N-Methylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals Product Code.: 11391537

N-Methylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11391537
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11391537

Brand: Thermo Scientific Alfa Aesar L02202.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-Methylhydroxylamine hydrochloride is used as an inorganic catalyst used in the transamidation of primary amides with amines. It is employed in constructing hydroxamates, important functional groups for the complexation of iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylhydroxylamine hydrochloride is used as an inorganic catalyst used in the transamidation of primary amides with amines. It is employed in constructing hydroxamates, important functional groups for the complexation of iron.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under dry inert gas. Protect from humidity and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4229-44-1
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.52
MDL Number MFCD00012597
InChI Key RGZRSLKIOCHTSI-UHFFFAOYSA-N
Synonym n-methylhydroxylamine hydrochloride, methylhydroxylammonium chloride, methylhydroxylamine hydrochloride, n-methylhydroxylamine hcl, unii-mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride, n-methylhydroxylammonium chloride, mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride 1:1, menhoh.hcl
PubChem CID 77906
IUPAC Name N-methylhydroxylamine;hydrochloride
SMILES [H+].[Cl-].CNO

Specifications

Melting Point 80°C to 86°C
Quantity 5 g
Beilstein 3541409
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 83.52
Percent Purity 98%
Chemical Name or Material N-Methylhydroxylamine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253455 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog NumberL02202.06,L02202.14
Lot Number10238417 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberL02202.06,L02202.14
Lot Number10147919 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02202.06,L02202.14
Lot Number10192353 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02202.06,L02202.14
Lot Number10156270 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL02202.06,L02202.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Methylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.