missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpiperidine, 99%, Thermo Scientific Chemicals

£15.08 - £163.82

Chemical Identifiers

CAS 626-67-5
Molecular Formula C6H14ClN
Molecular Weight (g/mol) 135.64
MDL Number MFCD00006491
InChI Key QDUXDCXILAPLAG-UHFFFAOYSA-N
Synonym n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
PubChem CID 12291
IUPAC Name 1-methylpiperidine
SMILES [H+].[Cl-].CN1CCCCC1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10001350
View Documents
Thermo Scientific Acros
127480100
10 mL Glass bottle
£15.08
10mL
Please to purchase this item. Need a web account? Register with us today!
10582521
View Documents Promotion Details
Thermo Scientific Acros
127482500
250 mL Glass bottle
£48.74
250mL
Please to purchase this item. Need a web account? Register with us today!
13428348
View Documents
Thermo Scientific Acros
127480010
1 L Glass bottle
£163.82
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

626-67-5
135.64
QDUXDCXILAPLAG-UHFFFAOYSA-N
12291
[H+].[Cl-].CN1CCCCC1
C6H14ClN
MFCD00006491
n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
1-methylpiperidine

Specifications

626-67-5
100.0
Colorless
105.0°C to 108.0°C
Authentic
C6H14ClN
MFCD00006491
0.82
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, acetone, toluene and
[H+].[Cl-].CN1CCCCC1
135.64
5 mPa.s (20°C)
99%
N-Methylpiperidine, 99%
98.5
-50.0°C
0.8200g/mL
3°C
98.5% min. (GC)
1.4368 to 1.4388
20,16
n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
QDUXDCXILAPLAG-UHFFFAOYSA-N
1-methylpiperidine
12291
99.18
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 210-959-9

missing translation for 'rtecsNumber' : TN1225000

missing translation for 'tsca' : TSCA

Product Identifier
  • N-Methylpiperidine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only