missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methylpropargylamine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Density | 0.8300g/mL |
Boiling Point | 82°C to 84°C |
Flash Point | 1°C |
Assay Percent Range | 98% min. (GC) |
Packaging | Glass bottle |
Linear Formula | HC∽CCH2NHCH3 |
Refractive Index | 1.4332 |
Quantity | 1 g |
Show More |
Safety and Handling
Product Identifier
- N-Methylpropargylamine
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : N-Methylpropargylamine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-Methylpropargylamine, 98+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction