missing translation for 'onlineSavingsMsg'
Learn More
N-Methylsulfanilic acid, 98+%, Thermo Scientific™ Product Code.: 11384414

N-Methylsulfanilic acid, 98+%, Thermo Scientific™

Product Code. 11384414
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11384414

Brand: Thermo Scientific Alfa Aesar L01016.06

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24447-99-2
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.213
MDL Number MFCD00014732
InChI Key QRAXZXPSAGQUNP-UHFFFAOYSA-N
Synonym 4-methylamino benzenesulfonic acid, 4-methylamino benzenesulphonic acid, n-methylsulphanilic acid, n-methylsulfanilic acid, benzenesulfonic acid,4-methylamino, n-methylaniline-p-sulfonic acid, maybridge1_004198, n-methylsulfanilic acid, 98+%
PubChem CID 90505
IUPAC Name 4-(methylamino)benzenesulfonic acid
SMILES CNC1=CC=C(C=C1)S(=O)(=O)O

Specifications

Melting Point ∽260°C (decomposition)
Quantity 5 g
Beilstein 2094129
Formula Weight 187.21
Percent Purity ≥98%
Chemical Name or Material N-Methylsulfanilic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Methylsulfanilic acid, 98+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.