Learn More
N-Methylthiourea, 97%, Thermo Scientific Chemicals
£33.80 - £108.00
Chemical Identifiers
CAS | 598-52-7 |
---|---|
Molecular Formula | C2H6N2S |
Molecular Weight (g/mol) | 90.14 |
MDL Number | MFCD00004938 |
InChI Key | KQJQICVXLJTWQD-UHFFFAOYSA-N |
Synonym | n-methylthiourea, 1-methylthiourea, 1-methyl-2-thiourea, methylthiocarbamide, thiourea, methyl, monomethylthiourea, 1-methylthiocarbamide, methyl thiourea, n-methylthiocarbamide, urea, 1-methyl-2-thio |
PubChem CID | 2723704 |
IUPAC Name | methylthiourea |
SMILES | CNC(=S)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10041320
|
Thermo Scientific Acros
127900250 |
25 g | Glass bottle |
£33.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10337010
|
Thermo Scientific Acros
127901000 |
100 g | Glass bottle |
£108.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
598-52-7 | |
90.14 | |
KQJQICVXLJTWQD-UHFFFAOYSA-N | |
2723704 | |
CNC(=S)N |
C2H6N2S | |
MFCD00004938 | |
n-methylthiourea, 1-methylthiourea, 1-methyl-2-thiourea, methylthiocarbamide, thiourea, methyl, monomethylthiourea, 1-methylthiocarbamide, methyl thiourea, n-methylthiocarbamide, urea, 1-methyl-2-thio | |
methylthiourea |
Specifications
598-52-7 | |
100.0 | |
White to Yellow | |
96% min. (HPLC) | |
CH3NHCSNH2 | |
04, 70 | |
Solubility in water: 165g/L (20°C). Other solubilities: 25g/L isopropanol (20°C),insoluble in non-polar solvents | |
CNC(=S)N | |
90.14 | |
90.14 | |
Crystalline Powder |
96.0 | |
118°C to 123°C | |
Authentic | |
C2H6N2S | |
MFCD00004938 | |
n-methylthiourea, 1-methylthiourea, 1-methyl-2-thiourea, methylthiocarbamide, thiourea, methyl, monomethylthiourea, 1-methylthiocarbamide, methyl thiourea, n-methylthiocarbamide, urea, 1-methyl-2-thio | |
KQJQICVXLJTWQD-UHFFFAOYSA-N | |
methylthiourea | |
2723704 | |
97% | |
N-Methylthiourea, 97% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Suspected of damaging the unborn child.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-936-6
missing translation for 'rtecsNumber' : YT9000000
- N-Methylthiourea
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Reproductive toxicity Category 2
Hazard Statement
- H361d-Suspected of damaging the unborn child.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only