missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(acryloyl)cystamine, 98%, Thermo Scientific Chemicals

Product Code. 11349618
Click to view available options
Quantity:
1 g
5 g
25 g

Used as a reversible cross-linker for polyacrylamide gels

N,N'-Bis(acryloyl)cystamine is used as a reversible cross-linker for polyacrylamide gels. It is also used for DNA purification. Further, it finds application in Suzuki reaction and fabrication of hydrogel modified plasmonic crystal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60984-57-8
Molecular Formula C10H16N2O2S2
Molecular Weight (g/mol) 260.37
MDL Number MFCD00036225
InChI Key DJVKJGIZQFBFGS-UHFFFAOYSA-N
Synonym n,n'-bis acryloyl cystamine, n,n'-bisacrylylcystamine, 2-propenamide, n,n'-dithiodi-2,1-ethanediyl bis, 2,2'-bisacrylamino diethyldisulfide, bis-acrylylcystamine: bac, n,n'-dithiodi-2,1-ethanediyl bis acrylamide, n-2-2-prop-2-enamido ethyl disulfanyl ethyl prop-2-enamide, n-2-2-prop-2-enoylamino ethyldisulfanyl ethyl prop-2-enamide, acmc-20ak9p, n,n-bis-acrylylcystamine
PubChem CID 100602
IUPAC Name N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide
SMILES C=CC(=O)NCCSSCCNC(=O)C=C

Specifications

Melting Point 123°C to 124°C
Assay Percent Range 98%
Quantity 5 g
Solubility Information Soluble in acetic acid. Slightly soluble in water.
Formula Weight 260.37
Physical Form Powder
Chemical Name or Material N, N′-Bis(acryloyl)cystamine
compliance-icons
Product Identifier
  • N,N'-Bis(acryloyl)cystamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N,N-(Dithiodi-2,1-ethanediyl)bis(acrylamide)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'