missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, 95%, Thermo Scientific Chemicals

Product Code. 15486926
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31687-58-8
Molecular Formula C28H48N4O6
Molecular Weight (g/mol) 536.714
InChI Key WBGMQHNUPJENDC-MERQFXBCSA-N
Synonym boc-his boc-oh.dcha, boc-his boc-oh dcha, boc-his boc-ohdcha, boc-his boc .dcha, n,1-bis tert-butoxycarbonyl-l-histidine, compound with dicyclohexylamine 1:1, pubchem18934, n,1-bis boc-l-histidine dicyclohexylamine salt, boc-his boc-ohcha, boc-his boc-oh?cha, boc-his boc-oh toluene solvate
PubChem CID 16211319
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]imidazol-4-yl]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(=O)OC(C)(C)C)C(=O)O.C1CCC(CC1)NC2CCCCC2

Specifications

Melting Point 160°C to 165°C (decomposition)
Color White
Formula Weight 536.7
Percent Purity 95%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N,N'-Di-Boc-L-histidine dicyclohexylammonium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'