missing translation for 'onlineSavingsMsg'
Learn More
N,N'-Di-n-butylthiourea, 98%, Thermo Scientific Chemicals Product Code.: 11442808

N,N'-Di-n-butylthiourea, 98%, Thermo Scientific Chemicals

Product Code. 11442808
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11442808

Brand: Thermo Scientific Alfa Aesar A16783.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N,N'-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-46-6
Molecular Formula C9H20N2S
Molecular Weight (g/mol) 188.333
MDL Number MFCD00004926
InChI Key KFFQABQEJATQAT-UHFFFAOYSA-N
Synonym 1,3-dibutyl-2-thiourea, n,n'-dibutylthiourea, thiourea, n,n'-dibutyl, pennzone b, n,n'-di-n-butylthiourea, thiate u, 1,3-di-n-butyl-2-thiourea, urea, 1,3-dibutyl-2-thio, usaf ek-2138, urea, 1,3-di-n-butyl-2-thioShow MoreShow Less
PubChem CID 2723622
IUPAC Name 1,3-dibutylthiourea
SMILES CCCCNC(=S)NCCCC

Specifications

Melting Point 64°C to 67°C
Odor Odorless
Quantity 250 g
Beilstein 507434
Solubility Information Insoluble in water.
Formula Weight 188.34
Percent Purity 98%
Chemical Name or Material N,N'-Di-n-butylthiourea

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253430 Certificate TypeCertificate of Analysis Date25/12/2024 Catalog NumberA16783.0B,A16783.18,A16783.30
Lot NumberB1094A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16783.0B,A16783.18,A16783.30
Lot Number10227485 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16783.0B,A16783.18,A16783.30
Lot Number10185973 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16783.0B,A16783.18,A16783.30
Lot NumberC4968C Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16783.0B,A16783.18,A16783.30
compliance-icons
Product Identifier
  • N,N'-Di-n-butylthiourea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Dibutyl thiourea

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N'-Di-n-butylthiourea, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.