missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di(o-tolyl)ethylenediamine, 98%, Thermo Scientific™

Product Code. 11441537
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-92-8
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00048073
InChI Key ZQMPWXFHAUDENN-UHFFFAOYSA-N
Synonym n,n'-di-o-tolylethylenediamine, n,n'-di o-tolyl ethylenediamine, n,n'-ethylenedi-o-toluidine, 1,2-ethanediamine, n,n'-bis 2-methylphenyl, 1,2-ethanediamine, n1,n2-bis 2-methylphenyl, n,n'-bis 2-methylphenyl ethane-1,2-diamine, 2-methylphenyl 2-2-methylphenyl amino ethyl amine, n1,n2-bis 2-methylphenyl ethane-1,2-diamine, cambridge id 5185196, n,n'-di-o-tolylethylendiamin
PubChem CID 66759
IUPAC Name N,N'-bis(2-methylphenyl)ethane-1,2-diamine
SMILES CC1=CC=CC=C1NCCNC1=CC=CC=C1C

Specifications

Melting Point 70°C to 73°C
Quantity 100 g
Formula Weight 240.35
Percent Purity 98%
Chemical Name or Material N,N'-Di(o-tolyl)ethylenediamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'