missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Di-tert-butylethylenediamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
N,N'-Di-tert-butylethylenediamine is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N′-Di-tert-butylethylenediamine is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.
Solubility
Soluble (33 g/L) (25°C),
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
N,N′-Di-tert-butylethylenediamine is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.
Solubility
Soluble (33 g/L) (25°C),
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
Specifications
Boiling Point | 132°C (100 mmHg) |
Flash Point | 62°C (143°F) |
Linear Formula | (CH3)3CNHCH2CH2NHC(CH3)3 |
Quantity | 5 g |
UN Number | UN2735 |
Beilstein | 1738432 |
Formula Weight | 172.32 |
Percent Purity | 98% |
Chemical Name or Material | N,N'-Di-tert-butylethylenediamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'