missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diaminoguanidine monohydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11495207
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36062-19-8
Molecular Formula CH8ClN5
Molecular Weight (g/mol) 125.56
MDL Number MFCD00012948
InChI Key HAZRIBSLCUYMQP-UHFFFAOYSA-N
Synonym 1,3-diaminoguanidine hydrochloride, unii-014y506oeg, 1,3-diaminoguanidine monohydrochloride, carbonimidic dihydrazide, hydrochloride, n,n'-diaminoguanidine monohydrochloride, n,n'-diaminoguanidine, carbonimidic dihydrazide, hydrochloride 1:?, 1,2-diaminoguanidine hydrochloride, n,n'-diaminoguandine, hydrochloride, pubchem20888
PubChem CID 9566041
IUPAC Name 1,2-diaminoguanidine;hydrochloride
SMILES C(=NN)(N)NN.Cl

Specifications

Melting Point ∼181°C (decomposition)
Quantity 5 g
Beilstein 3556702
Sensitivity Hygroscopic
Formula Weight 125.56
Percent Purity 98%
Chemical Name or Material N,N'-Diaminoguanidine monohydrochloride
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'