missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzylethylenediamine diacetate, 99%, Thermo Scientific Chemicals

Product Code. 11314865
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-75-8
Molecular Formula C20H28N2O4
Molecular Weight (g/mol) 360.454
MDL Number MFCD00040588
InChI Key MTRNNCLQPVCDLF-UHFFFAOYSA-N
Synonym n,n'-dibenzylethylenediamine diacetate, n1,n2-dibenzylethane-1,2-diamine diacetate, n,n'-dibenzyl ethylenediamine diacetate, benzathine diacetate, dbed diacetate, unii-x18b8x2c22, n,n'-dibenzylethylenediammonium di acetate, 1,2-ethanediamine, n,n'-bis phenylmethyl-, diacetate, bis acetic acid ; benzathine, 1,2-di benzylamino ethane diacetate
PubChem CID 31228
IUPAC Name acetic acid;N,N'-dibenzylethane-1,2-diamine
SMILES CC(=O)O.CC(=O)O.C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2

Specifications

Melting Point 117°C to 118°C
Quantity 100 g
Beilstein 3781362
Merck Index 14,1064
Formula Weight 360.46
Percent Purity 99%
Chemical Name or Material N,N'-Dibenzylethylenediamine diacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'