missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dibenzylhydroxylamine, 98%, Thermo Scientific Chemicals Product Code.: 11440607

N,N-Dibenzylhydroxylamine, 98%, Thermo Scientific Chemicals

Product Code. 11440607
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11440607

Brand: Thermo Scientific Alfa Aesar B22447.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-07-8
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00004772
InChI Key GXELTROTKVKZBQ-UHFFFAOYSA-N
Synonym dibenzylhydroxylamine, n,n-dibenzylhydroxyamine, benzenemethanamine, n-hydroxy-n-phenylmethyl, hydroxylamine, n,n-dibenzyl, bisbenzylhydroxylamine, n-benzyl-n-hydroxy-1-phenylmethanamine, acmc-1bivc, n.n-dibenzylhydroxylamine, maybridge3_004477, hydroxylamine,n-dibenzyl
PubChem CID 69297
IUPAC Name N,N-dibenzylhydroxylamine
SMILES ON(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Melting Point 120°C to 125°C
Quantity 100 g
Beilstein 978234
Formula Weight 213.28
Percent Purity 98%
Chemical Name or Material N,N-Dibenzylhydroxylamine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241224 Certificate TypeCertificate of Analysis Date10/06/2022 Catalog NumberB22447.22,B22447.14,B22447.06
Lot Number10241170 Certificate TypeCertificate of Analysis Date09/06/2022 Catalog NumberB22447.22,B22447.14,B22447.06
Lot Number10240965 Certificate TypeCertificate of Analysis Date02/06/2022 Catalog NumberB22447.22,B22447.14,B22447.06
Lot Number10194043 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22447.22,B22447.14,B22447.06
Lot Number10227123 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22447.22,B22447.14,B22447.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dibenzylhydroxylamine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.