missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

 

Brand:  Thermo Scientific Acros 429021000

Product Code. 10892621

  • £39.20 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

N, N'-Dicyclohexylcarbodiimide
15.81, 81.7
White to Brown
87°C
AcroSeal™ Glass Bottle
C6H11N=C=NC6H11
100 mL
1.24
Solubility in water: insoluble. Other solubilities: soluble in organic solvents
C1CCC(CC1)N=C=NC2CCCCC2
206.33
CHEBI:53090
Crystalline Powder
538-75-0, 75-09-2
18.3, 84.19
1.2400g/mL
Authentic
C13H22N2
MFCD00011659
15, 3107
dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl
QOSSAOTZNIDXMA-UHFFFAOYSA-N
N,N'-dicyclohexylmethanediimine
10868
206.33
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only