Learn More
N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N-Dicyclohexylcarbodiimide, 99%, C13H22N2, CAS Number-538-75-0, carbodiimide, dicyclohexyl, bis cyclohexyl carbodiimide, dcc, 1,3-dicyclohexylcarbodiimide, carbodicyclohexylimide, n,n'-dicyclohexylcarbodiimide, n,n-dicyclohexylcarbodiimide, dcci, dicyclohexylcarbodiimide, dccd, 1kg
£28.24 - £2674.31
Chemical Identifiers
CAS | 538-75-0 |
---|---|
Molecular Formula | C13H22N2 |
Molecular Weight (g/mol) | 206.33 |
MDL Number | MFCD00011659 |
InChI Key | QOSSAOTZNIDXMA-UHFFFAOYSA-N |
Synonym | dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl |
PubChem CID | 10868 |
ChEBI | CHEBI:53090 |
IUPAC Name | N,N'-dicyclohexylmethanediimine |
SMILES | C1CCC(CC1)N=C=NC2CCCCC2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10030880
|
Thermo Scientific Acros
113901000 |
100 g | Glass bottle |
£28.24
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10568290
|
Thermo Scientific Acros
113902500 |
250 g | Glass bottle |
£52.26
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10548290
|
Thermo Scientific Acros
113900010 |
1 kg | Glass bottle |
£158.79
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10498083
|
Thermo Scientific Acros
113900050 |
5 kg | Plastic drum |
£560.79
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11320370
|
Thermo Scientific Acros
113900250 |
25 kg | Plastic drum |
£2674.31
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
538-75-0 | |
206.33 | |
QOSSAOTZNIDXMA-UHFFFAOYSA-N | |
10868 | |
N,N'-dicyclohexylmethanediimine |
C13H22N2 | |
MFCD00011659 | |
dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl | |
CHEBI:53090 | |
C1CCC(CC1)N=C=NC2CCCCC2 |
Specifications
538-75-0 | |
100.0 | |
White to Yellow | |
>110°C | |
98.5% min. (GC) | |
C6H11N=C=NC6H11 | |
01,231; 02,126; 03,91; 04,141; 05,206; 06,174; 07,100; 08,162; 09,156; 10,142; 11,173; 13,333; 14,131; 15,131; 16,128 | |
dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl | |
QOSSAOTZNIDXMA-UHFFFAOYSA-N | |
N,N'-dicyclohexylmethanediimine | |
10868 | |
5.81 mPa.s (55°C) | |
99% | |
N,N′-Dicyclohexylcarbodiimide |
98.5 | |
33.0°C to 35.0°C | |
122.0°C to 124.0°C (6.0 mmHg) | |
Authentic | |
C13H22N2 | |
MFCD00011659 | |
15,3107 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents,100g/l dichloromethane | |
C1CCC(CC1)N=C=NC2CCCCC2 | |
206.33 | |
CHEBI:53090 | |
206.33 | |
Crystalline Mass or Waxy Solid |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-704-1
- N,N''-Dicyclohexylcarbodiimide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H317-May cause an allergic skin reaction.
- H310-Fatal in contact with skin.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only