missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-2,2,2-trifluoroacetamide, 97%, Thermo Scientific™

Product Code. 15550624
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 360-92-9
Molecular Formula C6H10F3NO
Molecular Weight (g/mol) 169.15
InChI Key CODXZFSZJFCVBE-UHFFFAOYSA-N
Synonym n,n-diethyltrifluoroacetamide, acetamide, n,n-diethyl-2,2,2-trifluoro, acmc-1cmeq, diethyl trifluoroacetamide, n,n-diethyltrifluoro-acetamide, trifluoroacetic acid diethylamide, n,n-diethyl-2,2,2-trifluoroacetamide, n,n-diethyl-2,2,2-tris fluoranyl ethanamide
PubChem CID 225443
IUPAC Name N,N-diethyl-2,2,2-trifluoroacetamide
SMILES CCN(CC)C(=O)C(F)(F)F

Specifications

Density 1.1420g/mL
Boiling Point 65.0°C to 67.0°C (24.0 mmHg)
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula F3CC(=O)N(CH2CH3)2
Quantity 5 g
Specific Gravity 1.142
Solubility Information Solubility in water: immiscible.
Formula Weight 169.15
Percent Purity 97%
Chemical Name or Material N,N-Diethyl-2,2,2-trifluoroacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Diethyl-2,2,2-trifluoroacetamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'