missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethyl-3-methylbenzamide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.
Solubility
Soluble in water.
Notes
Keep away from strong oxidizing agents, strong acids, strong bases, strong reducing agents. Keep ate room temperature.
It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.
Solubility
Soluble in water.
Notes
Keep away from strong oxidizing agents, strong acids, strong bases, strong reducing agents. Keep ate room temperature.
Specifications
Density | 0.997 |
Boiling Point | 110°C to 112°C (1 mmHg) |
Flash Point | 145°C (293°F) |
Refractive Index | 1.523 |
Quantity | 100 g |
Beilstein | 2046711 |
Merck Index | 14,2856 |
Solubility Information | Soluble in water. |
Formula Weight | 191.27 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'