missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-4-methylbenzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15421178
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15421178

Brand: Thermo Scientific Alfa Aesar H55053.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 649-15-0
Molecular Formula C11H17NO2S
Molecular Weight (g/mol) 227.322
MDL Number MFCD00059369
InChI Key AOJBACHWNDMRQP-UHFFFAOYSA-N
Synonym n,n-diethyl-p-toluenesulfonamide, p-toluenesulfonamide, n,n-diethyl, p-toluenesulfonyl-n,n-diethylamide, benzenesulfonamide, n,n-diethyl-4-methyl, p-toluenesulfonyl-n-diethylamide, n,n-diethyl-4-methylbenzolsulfonamid, acmc-209nol, maybridge1_002134, cambridge id 5116375, p-toluenesulfonamide,n-diethyl
PubChem CID 69539
IUPAC Name N,N-diethyl-4-methylbenzenesulfonamide
SMILES CCN(CC)S(=O)(=O)C1=CC=C(C=C1)C

Specifications

Melting Point 60°C
Color White
Quantity 1 g
Formula Weight 227.32
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material N,N-Diethyl-4-methylbenzenesulfonamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.