Learn More
N,N-Diethyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals
£18.29 - £297.48
Chemical Identifiers
CAS | 93-05-0 |
---|---|
MDL Number | MFCD00007861 |
InChI Key | QNGVNLMMEQUVQK-UHFFFAOYSA-N |
Synonym | n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline |
PubChem CID | 7120 |
IUPAC Name | 4-N,4-N-diethylbenzene-1,4-diamine |
SMILES | CCN(CC)C1=CC=C(C=C1)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10025710
|
Thermo Scientific Acros
407390050 |
5 g | Glass bottle |
£18.29
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10206061
|
Thermo Scientific Acros
407390250 |
25 g | Glass bottle |
£30.25
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10435675
|
Thermo Scientific Acros
407391000 |
100 g | Glass bottle |
£80.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10741325
|
Thermo Scientific Acros
407395000 |
500 g | Glass bottle |
£297.48
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93-05-0 | |
QNGVNLMMEQUVQK-UHFFFAOYSA-N | |
7120 | |
CCN(CC)C1=CC=C(C=C1)N |
MFCD00007861 | |
n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline | |
4-N,4-N-diethylbenzene-1,4-diamine |
Specifications
93-05-0 | |
100.0 | |
0.9880g/mL | |
139°C | |
97.5% min. (GC) | |
(CH3CH2)2NC6H4NH2 | |
MFCD00007861 | |
n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline | |
QNGVNLMMEQUVQK-UHFFFAOYSA-N | |
4-N,4-N-diethylbenzene-1,4-diamine | |
7120 | |
98% | |
N, N-Diethyl-p-phenylenediamine |
97.5 | |
23.0°C | |
268.0°C | |
Authentic | |
C10H16N2 | |
1.5690 to 1.5730 (20°C, 589nm) | |
0.988 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
CCN(CC)C1=CC=C(C=C1)N | |
164.25 | |
164.25 | |
Liquid or Low Melting Solid |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-214-1
- N,N-Diethyl-p-phenylenediamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only