missing translation for 'onlineSavingsMsg'
Learn More
N,N-Diethyl-p-phenylenediamine sulfate, 97%, Thermo Scientific Chemicals Product Code.: 11413428

N,N-Diethyl-p-phenylenediamine sulfate, 97%, Thermo Scientific Chemicals

Product Code. 11413428
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11413428

Brand: Thermo Scientific Alfa Aesar A17832.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N,N-Diethyl-p-phenylenediamine sulfate is used for the spectrophotometric determination of drugs such as salbutamol sulphate, ritodrine hydrochloride and isoxsuprine hydrochloride containing phenolic group. It is also used in the analysis of drugs like dapsone hydrochloride, sulfamethoxazole and sulfadiazine containing aromatic amino group. Further, it is used as a color developer and film developer in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethyl-p-phenylenediamine sulfate is used for the spectrophotometric determination of drugs such as salbutamol sulphate, ritodrine hydrochloride and isoxsuprine hydrochloride containing phenolic group. It is also used in the analysis of drugs like dapsone hydrochloride, sulfamethoxazole and sulfadiazine containing aromatic amino group. Further, it is used as a color developer and film developer in photography.

Solubility
Soluble in water and alcohol.

Notes
Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6283-63-2
Molecular Formula C10H18N2O4S
Molecular Weight (g/mol) 262.324
MDL Number MFCD00012993
InChI Key AYLDJQABCMPYEN-UHFFFAOYSA-N
Synonym n,n-diethyl-p-phenylenediamine sulfate, n1,n1-diethylbenzene-1,4-diamine sulfate, 4-amino-n,n-diethylaniline sulfate, diethyl-p-phenylenediamine sulfate, n,n-diethyl-1,4-phenylenediamine sulfate, unii-usp19t3gda, 1,4-benzenediamine, n,n-diethyl-, sulfate 1:1, 1,4-benzenediamine, n,n-diethyl-, sulfate, n,n-diethyl-1,4-benzenediamine sulfate, p-phenylenediamine, n,n-diethyl-, sulfate 1:1
PubChem CID 80166
IUPAC Name 4-N,4-N-diethylbenzene-1,4-diamine;sulfuric acid
SMILES CCN(CC)C1=CC=C(C=C1)N.OS(=O)(=O)O

Specifications

Melting Point 183°C to 188°C
Quantity 250 g
Beilstein 4219768
Sensitivity Light sensitive
Solubility Information Soluble in water and alcohol.
Formula Weight 262.33
Percent Purity 97%
Chemical Name or Material N,N-Diethyl-p-phenylenediamine sulfate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Diethyl-p-phenylenediamine sulfate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.