missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylaniline, 99%, Thermo Scientific Chemicals

Product Code. 11413204
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.

Solubility
Miscible with water and acetone. Slightly miscible with chloroform, alcohol and ether.      

Notes
Incompatible with strong oxidizing agents and strong acids.                  .
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-66-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00009042,MFCD31699978
InChI Key GGSUCNLOZRCGPQ-UHFFFAOYSA-N
Synonym diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine
PubChem CID 7061
IUPAC Name N,N-diethylaniline
SMILES CCN(CC)C1=CC=CC=C1

Specifications

Melting Point -38°C
Density 0.936
Boiling Point 215°C to 216°C
Flash Point 85°C (185°F)
Refractive Index 1.542
Quantity 500 mL
UN Number UN2432
Beilstein 742483
Merck Index 14,3114
Solubility Information Miscible with water and acetone. Slightly miscible with chloroform,alcohol and ether.
Formula Weight 149.24
Percent Purity 99%
Chemical Name or Material N,N-Diethylaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.