Learn More
Chemical Identifiers
CAS | 91-66-7 |
---|---|
Molecular Formula | C10H15N |
Molecular Weight (g/mol) | 149.24 |
MDL Number | MFCD00009042,MFCD31699978 |
InChI Key | GGSUCNLOZRCGPQ-UHFFFAOYSA-N |
Synonym | diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine |
PubChem CID | 7061 |
IUPAC Name | N,N-diethylaniline |
SMILES | CCN(CC)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10477940
|
Thermo Scientific Acros
181062500 |
250 mL | Glass bottle |
£18.89
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10032360
|
Thermo Scientific Acros
181060010 |
1 L | Glass bottle |
£46.03
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10154390
|
Thermo Scientific Acros
181060025 |
2.5 L | Glass bottle |
£86.33
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
91-66-7 | |
149.24 | |
GGSUCNLOZRCGPQ-UHFFFAOYSA-N | |
7061 | |
CCN(CC)C1=CC=CC=C1 |
C10H15N | |
MFCD00009042,MFCD31699978 | |
diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine | |
N,N-diethylaniline |
Specifications
91-66-7 | |
100.0 | |
Yellow | |
215.0°C to 217.0°C | |
Authentic | |
C10H15N | |
1.5410 to 1.5430 | |
12, 164 | |
15, 3125 | |
diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine | |
GGSUCNLOZRCGPQ-UHFFFAOYSA-N | |
N,N-diethylaniline | |
7061 | |
99% | |
N, N-Diethylaniline, 99% |
98.5 | |
-38.0°C | |
0.9330g/mL | |
84°C | |
99% | |
C6H5N(C2H5)2 | |
MFCD00009042,MFCD31699978 | |
05,212; 16,182 | |
0.933 | |
Solubility in water: 10-27g/L. Other solubilities: slightly soluble in alcohol,chloroform and ether | |
CCN(CC)C1=CC=CC=C1 | |
149.24 | |
149.24 | |
Liquid |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-088-8
missing translation for 'rtecsNumber' : BX3400000
missing translation for 'tsca' : TSCA
- N,N-Diethylaniline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : N,N-Diethylaniline
RUO – Research Use Only